Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorbitan monostearate

Supplier: Thermo Scientific Chemicals
Synonyms: Sorbitan monooctadecanoate(2R,3R,4R,5S)-Hexane-1,2,3,4,5,6-hexaol monooctadecanoateSorbitan stearate

L11435-30 L11435-0B L11435-0B L11435-30
AAAL11435-0BEA 84.39 USD InStock
CAAAAL11435-0B AAAL11435-0B CAAAAL11435-30 AAAL11435-30
Sorbitan monostearate
Sorbitan monostearate
Emulsifier used in cosmetics, pharmaceuticals and food preparation


Formula: C₂₄H₄₆O₆
MW: 430.63 g/mol
Melting Pt: 53…57 °C
Density: 1.00
Flash Pt: >149 °C(300 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005366
CAS Number: 1338-41-6
EINECS: 215-664-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty