Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Ethoxyphenyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 1-Ethoxy-4-isocyanatobenzene

L11509-06 L11509-06 L11509-14 L11509-14
AAAL11509-14EA 128.99 USD InStock
CAAAAL11509-06 AAAL11509-14 CAAAAL11509-14 AAAL11509-06
4-Ethoxyphenyl isocyanate 97%
4-Ethoxyphenyl isocyanate


Formula: C₉H₉NO₂
MW: 163.18 g/mol
Boiling Pt: 235 °C
Density: 1.110
Flash Pt: 105 °C(221 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00013877
CAS Number: 32459-62-4
UN: 2206
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty