Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hexyl methacrylate 97% stabilized

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Methacrylic Acid Hexyl Ester

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 100 ppm

L11569-22 L11569-14 L11569-22 L11569-14
AAAL11569-14EA 33.59 USD InStock
AAAL11569-14 AAAL11569-22 CAAAAL11569-14 CAAAAL11569-22
Hexyl methacrylate 97% stabilized
Hexyl methacrylate


Formula: C₁₀H₁₈O₂
MW: 170.25 g/mol
Boiling Pt: 204 °C
Melting Pt: –57 °C
Density: 0.885
Flash Pt: 80 °C(176 °F)
Storage Temperature: Ambient
MDL Number: MFCD00015283
CAS Number: 142-09-6
EINECS: 205-521-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty