Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

S-Benzyl-L-cysteine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: H-Cys(Bzl)-OH

L11666-14 L11666-06 L11666-06 L11666-14
AAAL11666-14EA 53.39 USD InStock
CAAAAL11666-06 AAAL11666-14 CAAAAL11666-14 AAAL11666-06
S-Benzyl-L-cysteine 99%
S-Benzyl-L-cysteine


Formula: C₁₀H₁₃NO₂S
MW: 211.28 g/mol
Melting Pt: ca 214 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00002613
CAS Number: 3054-01-1
EINECS: 221-273-4
Order Now
Specification Test Results
Optical rotation +28° (c=1.5 in 1N NaOH)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty