Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Chloro-2,4-dinitrotoluene 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 1-Chloro-5-methyl-2,4-dinitrobenzene

L11808-14 L11808-14 L11808-06 L11808-06
AAAL11808-14EA 241.99 USD InStock
AAAL11808-14 CAAAAL11808-06 CAAAAL11808-14 AAAL11808-06
5-Chloro-2,4-dinitrotoluene 97%
5-Chloro-2,4-dinitrotoluene


Formula: C₇H₅ClN₂O₄
MW: 216.58 g/mol
Melting Pt: 89…91 °C
Storage Temperature: Ambient
MDL Number: MFCD00041235
CAS Number: 51676-74-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty