Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-alanine 96%

Supplier: Thermo Scientific Chemicals
Synonyms: Acetyl-L-alanineAc-Ala-OH

L11811-03 L11811-06 L11811-06 L11811-03
AAAL11811-06EA 80.06 USD InStock
AAAL11811-06 CAAAAL11811-06 AAAL11811-03 CAAAAL11811-03
N-Acetyl-L-alanine 96%
N-Acetyl-L-alanine


Formula: C₅H₉NO₃
MW: 131.131 g/mol
Melting Pt: 122…128 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00063132
CAS Number: 97-69-8
EINECS: 202-602-0
Order Now
Specification Test Results
Optical rotation -64° (c=2 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty