Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA dipotassium salt dihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Ethylenedinitrilotetra-acetic acid dipotassium salt dihydrateEdetic acid dipotassium salt dihydrateDipotassium dihydrogen ethylenediaminetetraacetate dihydratePotassium 2,2'-((2-(bis(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrateEthylenediaminetetra-acetic acid dipotassium salt dihydrate

L11842-22 L11842-36 L11842-22 L11842-36
AAAL11842-22EA 70.25 USD InStock
CAAAAL11842-22 CAAAAL11842-36 AAAL11842-22 AAAL11842-36
EDTA dipotassium salt dihydrate 99%
EDTA dipotassium salt dihydrate


Formula: C₁₀H₁₄K₂N₂O₈·2H₂O
MW: 404.46 g/mol
Melting Pt: ca 272 °C (dec.)
Density: 1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150036
CAS Number: 25102-12-9
EINECS: 217-895-0
Merck Index: 13,03546
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty