Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Methoxyethyl acrylate 98% stabilized

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (2-Methoxyethyl) acrylateEthylene Glycol Monomethyl Ether AcrylateAcrylic Acid 2-Methoxyethyl EsterEthylene glycol methyl ether acrylate

Stabilization: Stabilised with 4-methoxyphenol (MEHQ) 50 ppm-100 ppm

L11902-18 L11902-30 L11902-30 L11902-18 L11902-0B L11902-0B
AAAL11902-0BEA 125.99 USD InStock
AAAL11902-0B CAAAAL11902-0B AAAL11902-30 CAAAAL11902-30 AAAL11902-18 CAAAAL11902-18
2-Methoxyethyl acrylate 98% stabilized
2-Methoxyethyl acrylate


Formula: C₆H₁₀O₃
MW: 130.14 g/mol
Boiling Pt: 162 °C
Melting Pt: <lt/>–30 °C
Density: 1.008
Flash Pt: 56 °C(133 °F)
Storage Temperature: Ambient
MDL Number: MFCD00048149
CAS Number: 3121-61-7
EINECS: 221-499-3
UN: 1992
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty