Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

EDTA tripotassium salt dihydrate 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Tripotassium hydrogen ethylenediaminetetraacetate dihydrateEdetic acid tripotassium salt dihydratePotassium 2,2'-((2-((carboxylatomethyl)(carboxymethyl)amino)ethyl)azanediyl)diacetate dihydrateEthylenedinitrilotetra-acetic acid tripotassium salt dihydrateEthylenediaminetetra-acetic acid tripotassium salt dihydrate

L11991-30 L11991-18 L11991-30 L11991-18
AAAL11991-18EA 48.49 USD InStock
CAAAAL11991-18 AAAL11991-18 AAAL11991-30 CAAAAL11991-30
EDTA tripotassium salt dihydrate 99%
EDTA tripotassium salt dihydrate


Formula: C₁₀H₁₃K₃N₂O₈·2H₂O
MW: 442.55 g/mol
Melting Pt: ca 182 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00149672
CAS Number: 65501-24-8
EINECS: 241-543-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty