Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3,5-Dichlorobenzylamine 94%

Supplier: Thermo Scientific Chemicals
Danger

L12054-06 L12054-06
AAAL12054-06EA 278.99 USD InStock
AAAL12054-06 CAAAAL12054-06
3,5-Dichlorobenzylamine 94%
3,5-Dichlorobenzylamine


Formula: C₇H₇Cl₂N
MW: 176.04 g/mol
Boiling Pt: 141…144 °C/15 mm
Density: 1.320
Storage Temperature: Ambient
MDL Number: MFCD00052681
CAS Number: 39989-43-0
EINECS: 000-000-0
UN: 2735
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty