Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sorbitan monolaurate

Supplier: Thermo Scientific Chemicals
Synonyms: Sorbitan laurateSorbitan monododecanoateSpan 20

L12099-0B L12099-30 L12099-0B L12099-30
AAAL12099-0BEA 104.48 USD InStock
AAAL12099-0B CAAAAL12099-0B AAAL12099-30 CAAAAL12099-30
Sorbitan monolaurate
Sorbitan monolaurate


Formula: C₁₈H₃₄O₆
MW: 346.46 g/mol
Density: 1.032
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00005365
CAS Number: 1338-39-2
EINECS: 215-663-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty