Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hexyldecanoic acid 97%

Supplier: Thermo Scientific Chemicals
Synonyms: Isopalmitic AcidPentadecane-7-carboxylic Acid

L12238-22 L12238-22 L12238-14 L12238-14
AAAL12238-14EA 30.52 USD InStock
AAAL12238-14 CAAAAL12238-14 AAAL12238-22 CAAAAL12238-22
2-Hexyldecanoic acid 97%
2-Hexyldecanoic acid


Formula: C₁₆H₃₂O₂
MW: 256.43 g/mol
Boiling Pt: 165…168 °C/2 mm
Melting Pt: 10 °C
Density: 0.880
Flash Pt: 174 °C(345 °F)
Storage Temperature: Ambient
MDL Number: MFCD00059651
CAS Number: 25354-97-6
EINECS: 246-885-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty