Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-1-(1-Naphthyl)ethyl isocyanate 97%

Supplier: Thermo Scientific Chemicals
Danger

L12286-MD L12286-03 L12286-MD L12286-03
AAAL12286-MDEA 41.99 USD InStock
AAAL12286-MD AAAL12286-03 CAAAAL12286-MD CAAAAL12286-03
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate 97%
(R)-(-)-1-(1-Naphthyl)ethyl isocyanate


Formula: C₁₃H₁₁NO
Boiling Pt: 106…108°/0.16m
Density: 1.118
Flash Pt: 93 °C(199 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00004015
CAS Number: 42340-98-7
EINECS: 255-759-2
UN: 2206
ADR: "6.1",III
Order Now
Specification Test Results
Optical rotation -47° (c=3.5 in toluene)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty