Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Nitro-5-(trifluoromethyl)aniline 97%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 3-Amino-4-nitrobenzotrifluoride

L12310-06 L12310-06 L12310-03 L12310-03
AAAL12310-06EA 288.99 USD InStock
AAAL12310-06 CAAAAL12310-03 AAAL12310-03 CAAAAL12310-06
2-Nitro-5-(trifluoromethyl)aniline 97%
2-Nitro-5-(trifluoromethyl)aniline


Formula: C₇H₅F₃N₂O₂
MW: 206.13
Melting Pt: 98…102 °C
Storage Temperature: Ambient
MDL Number: MFCD00042447
EINECS: 000-000-0
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty