Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-(2,2,2-Trifluoroacetyl)thiophene 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-(Trifluoroacetyl)thiophene

L12585-03 L12585-03 L12585-06 L12585-06
AAAL12585-03EA 36.09 USD InStock
AAAL12585-03 CAAAAL12585-06 CAAAAL12585-03 AAAL12585-06
2-(2,2,2-Trifluoroacetyl)thiophene 98%
2-(2,2,2-Trifluoroacetyl)thiophene


Formula: C₆H₅ClOS
MW: 180.15
Boiling Pt: 163…165 °C
Density: 1.403
Flash Pt: 58 °C(136 °F)
Storage Temperature: Ambient
MDL Number: MFCD00041427
UN: 1993
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty