Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2,4,6-Trimethylphenyl isocyanate 98%

Supplier: Thermo Scientific Chemicals
Danger

L12611-06 L12611-03 L12611-03 L12611-06
AAAL12611-03EA 30.39 USD InStock
CAAAAL12611-06 AAAL12611-03 CAAAAL12611-03 AAAL12611-06
2,4,6-Trimethylphenyl isocyanate 98%
2,4,6-Trimethylphenyl isocyanate


Formula: C₁₀H₁₁NO
Boiling Pt: 218…220 °C
Melting Pt: 43…46 °C
Flash Pt: 104 °C(219 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00019909
CAS Number: 2958-62-5
UN: 2811
ADR: "6.1",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty