Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Nitro-L-phenylalanine monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (S)-2-Amino-3-(4-nitrophenyl)propanoic acid monohydrate(S)-4-Nitrophenylalanine monohydrateH-Phe(4-NO2)-OH.H2O

L12709-06 L12709-03 L12709-06 L12709-03
AAAL12709-03EA 42.99 USD InStock
AAAL12709-03 AAAL12709-06 CAAAAL12709-06 CAAAAL12709-03
4-Nitro-L-phenylalanine monohydrate 98%
4-Nitro-L-phenylalanine monohydrate


Formula: C₉H₁₂N₂O₅
MW: 228.20 g/mol
Melting Pt: 236…239 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00051221
CAS Number: 207591-86-4
EINECS: 213-446-8
Order Now
Specification Test Results
Optical rotation +6.5° (c=1 in 5N HCl)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty