Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Amygdalin hydrate 96%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Amygdalin hydrateAmygdaloside hydrateLaetrile hydrateD-Mandelonitrile 6-O-β-D-glucosido-β-D-glucoside hydrateD-Mandelonitrile-β-gentiobioside hydrate

L12731-04 L12731-09 L12731-04 L12731-09
AAAL12731-09EA 78.39 USD InStock
CAAAAL12731-04 AAAL12731-09 AAAL12731-04 CAAAAL12731-09
D-Amygdalin hydrate 96%
D-Amygdalin hydrate
A benzylic glycoside that has been used as an antineoplastic agent


Formula: C₂₀H₂₇NO₁₁·xH₂O
MW: 457.44 g/mol (anhy)
Melting Pt: 215…220 °C
Storage Temperature: Ambient
MDL Number: MFCD00006598
CAS Number: 29883-15-6
EINECS: 249-925-3
Order Now
Specification Test Results
Appearance (Color) White
Form Crystalline solid
Assay (HPLC) 95.0%
Loss on Drying 10.0% @ 110°C
Optical Rotation -38 ±2° (c=1.2 in water) on dry wt. basis

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty