Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-2-Phenylbutyric acid 99%

Supplier: Thermo Scientific Chemicals
Warning

L12754-03 L12754-06 L12754-06 L12754-03
AAAL12754-03EA 87.59 USD InStock
CAAAAL12754-06 CAAAAL12754-03 AAAL12754-03 AAAL12754-06
(S)-(+)-2-Phenylbutyric acid 99%
(S)-(+)-2-Phenylbutyric acid


Formula: C₁₀H₁₂O₂
MW: 164.2 g/mol
Boiling Pt: 270…272 °C
Density: 1,055
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00063164
CAS Number: 4286-15-1
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty