Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Hexenoic acid 99%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 5-Hexenoicacid

L12863-06 L12863-03 L12863-06 L12863-03
AAAL12863-03EA 57.39 USD InStock
CAAAAL12863-03 AAAL12863-03 CAAAAL12863-06 AAAL12863-06
5-Hexenoic acid 99%
5-Hexenoic acid


Formula: C₆H₁₀O₂
MW: 114.14 g/mol
Boiling Pt: 107 °C/17 mm
Melting Pt: –37 °C
Density: 0.961
Flash Pt: 104 °C(219 °F)
Storage Temperature: Ambient
MDL Number: MFCD00046558
CAS Number: 1577-22-6
UN: 3265
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty