Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, Technical Grade

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Hydroxy-4-methoxybenzophenone-5-sulfonic Acid Hydrate2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid Hydrate

L12912-09 L12912-09 L12912-18 L12912-18
AAAL12912-18EA 270.99 USD InStock
CAAAAL12912-09 AAAL12912-18 CAAAAL12912-18 AAAL12912-09
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate tech. 85% may cont. up to 10% 2-propanol, Technical Grade
2-Hydroxy-4-methoxy-5-sulfobenzophenone hydrate


Formula: C₁₄H₁₂O₆S·H₂O
MW: 308.31 g/mol (anhy)
Melting Pt: 108…112 °C
Storage Temperature: Ambient
MDL Number: MFCD00024962
CAS Number: 4065-45-6
EINECS: 223-772-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty