Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Butyric acid 99+%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: n-Butyric acidButanoic acid

L13189-0F L13189-AP L13189-AP L13189-0F L13189-AE L13189-AE
AAAL13189-APEA 38.49 USD InStock
AAAL13189-AP CAAAAL13189-AE CAAAAL13189-0F AAAL13189-0F AAAL13189-AE CAAAAL13189-AP
Butyric acid 99+%
Butyric acid


Formula: C₄H₈O₂
MW: 88.11 g/mol
Boiling Pt: 162…164 °C
Melting Pt: –8 °C
Density: 0.964
Flash Pt: 69 °C(156 °F)
Storage Temperature: Ambient
MDL Number: MFCD00002814
CAS Number: 107-92-6
EINECS: 203-532-3
UN: 2820
ADR: "8",III
Merck Index: 13,01593
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty