Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(+)-Di-p-toluoyl-D-tartaric acid 98%

Supplier: Thermo Scientific Chemicals
Synonyms: (2S,3S)-(+)-Di-O-4-Toluoyl-D-Tartaric AcidDi-p-toluoyl-D-tartaric AcidDi-p-toluoyl-D-tartaric acid

L13223-22 L13223-06 L13223-14 L13223-22 L13223-14 L13223-06
AAAL13223-22EA 279.99 USD InStock
CAAAAL13223-22 AAAL13223-22 AAAL13223-06 CAAAAL13223-14 CAAAAL13223-06 AAAL13223-14
(+)-Di-p-toluoyl-D-tartaric acid 98%
(+)-Di-p-toluoyl-D-tartaric acid


Formula: C₂₀H₁₈O₈
MW: 386.36 g/mol
Melting Pt: 169…171 °C
Storage Temperature: Ambient
MDL Number: MFCD00008552
CAS Number: 32634-68-7
EINECS: 251-132-2
Order Now
Specification Test Results
Optical rotation +139° (c=1 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty