Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

L(+)-Lactic acid, anhydrous 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (S)-(+)-2-Hydroxypropionic acid(S)-2-Hydroxypropanoic acid(S)-(+)-Lactic acid(S)-(+)-2-Hydroxypropanoic acid

L13242-22 L13242-22 L13242-06 L13242-06 L13242-14 L13242-14
AAAL13242-06EA 94.37 USD InStock
AAAL13242-06 CAAAAL13242-22 AAAL13242-22 CAAAAL13242-14 AAAL13242-14 CAAAAL13242-06
L(+)-Lactic acid, anhydrous 98%
L(+)-Lactic acid
Useful chiral synthon, building block for depsipeptides


Formula: C₃H₆O₃
MW: 90.08 g/mol
Boiling Pt: 119 °C/12 mm
Melting Pt: 52…54 °C
Density: 1.22
Flash Pt: >110 °C(230 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00064266
CAS Number: 79-33-4
EINECS: 201-196-2
Merck Index: 13,05352
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty