Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1-Boc-piperazine 99%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: tert-butyl piperazine-1-carboxylatetert-Butyl 1-piperazinecarboxylate

L13363-03 L13363-03 L13363-06 L13363-06 L13363-14 L13363-14
AAAL13363-06EA 117 USD InStock
AAAL13363-06 CAAAAL13363-03 CAAAAL13363-14 CAAAAL13363-06 AAAL13363-03 AAAL13363-14
1-Boc-piperazine 99%
1-Boc-piperazine


Formula: C₉H₁₈N₂O₂
MW: 186.25 g/mol
Melting Pt: 44…48 °C
Storage Temperature: Ambient
MDL Number: MFCD00075265
CAS Number: 57260-71-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty