Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Hexahydro-p-xylylenediamine1,4-Di(aminomethyl)cyclohexane

L13407-14 L13407-06 L13407-03 L13407-03 L13407-14 L13407-06
AAAL13407-06EA 97.17 USD InStock
AAAL13407-06 CAAAAL13407-03 CAAAAL13407-14 AAAL13407-14 AAAL13407-03 CAAAAL13407-06
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture) 96%
1,4-Bis(aminomethyl)cyclohexane (cis and trans mixture)


Formula: C₈H₁₈N₂
MW: 142.25 g/mol
Boiling Pt: 239…244 °C
Density: 0.950
Flash Pt: 106 °C(222 °F)
Storage Temperature: Ambient
MDL Number: MFCD00039463
CAS Number: 2549-93-1
EINECS: 219-840-6
UN: 3267
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty