Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Bromophthalic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L13418-06 L13418-03 L13418-06 L13418-03
AAAL13418-03EA 51.39 USD InStock
CAAAAL13418-06 AAAL13418-03 CAAAAL13418-03 AAAL13418-06
4-Bromophthalic acid 98%
4-Bromophthalic acid


Formula: C₈H₅BrO₄
MW: 245.03 g/mol
Melting Pt: 162…164 °C
Storage Temperature: Ambient
MDL Number: MFCD00043538
CAS Number: 6968-28-1
EINECS: 230-183-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty