Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Bromo-3-pyridineacetic acid ≥98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 5-Bromo-3-pyridylacetic acid

L13579-06 L13579-03 L13579-03 L13579-06
AAAL13579-06EA 427.99 USD InStock
AAAL13579-06 CAAAAL13579-03 CAAAAL13579-06 AAAL13579-03
5-Bromo-3-pyridineacetic acid ≥98%
5-Bromo-3-pyridineacetic acid


Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
Melting Pt: 182…185 °C
Storage Temperature: Ambient
MDL Number: MFCD00829308
CAS Number: 39891-12-8
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty