Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

5-Acetylthiophene-2-carboxylic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L13588-14 L13588-06 L13588-14 L13588-06
AAAL13588-06EA 78.19 USD InStock
AAAL13588-06 AAAL13588-14 CAAAAL13588-06 CAAAAL13588-14
5-Acetylthiophene-2-carboxylic acid 98+%
5-Acetylthiophene-2-carboxylic acid


Formula: C₇H₆O₃S
MW: 170.19 g/mol
Melting Pt: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD00055512
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty