Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

O-Benzylhydroxylamine 96%

Supplier: Thermo Scientific Chemicals
Warning

L13601-06 L13601-06 L13601-14 L13601-14
AAAL13601-06EA 74.48 USD InStock
AAAL13601-06 CAAAAL13601-06 AAAL13601-14 CAAAAL13601-14
O-Benzylhydroxylamine 96%
O-Benzylhydroxylamine


Formula: C₇H₉NO
Boiling Pt: 118…119 °C/30 mm
Density: 1.058
Storage Temperature: Ambient
MDL Number: MFCD00221709
CAS Number: 622-33-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty