Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-leucine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-Leu-OH

L13926-14 L13926-06 L13926-06 L13926-14
AAAL13926-14EA 106.99 USD InStock
CAAAAL13926-14 AAAL13926-14 CAAAAL13926-06 AAAL13926-06
N-Acetyl-L-leucine 99%
N-Acetyl-L-leucine


Formula: C₈H₁₅NO₃
MW: 173.2 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD00065131
CAS Number: 1188-21-2
EINECS: 214-706-3
Order Now
Specification Test Results
Optical rotation -23° (c=5 in ethanol)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty