Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tetrafluoroboric acid ∼50% (w/w) in aqueous solution

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Fluoboric acid

L14037-22 L14037-36 L14037-0E L14037-36 L14037-0E L14037-22
AAAL14037-22EA 57.86 USD InStock
AAAL14037-22 CAAAAL14037-22 CAAAAL14037-0E CAAAAL14037-36 AAAL14037-36 AAAL14037-0E
Tetrafluoroboric acid ∼50% (w/w) in aqueous solution
Tetrafluoroboric acid


Formula: HBF₄
MW: 87.81 g/mol
Boiling Pt: 130 °C (dec.)
Density: 1.4
Storage Temperature: Ambient
MDL Number: MFCD00011345
CAS Number: 16872-11-0
EINECS: 240-898-3
UN: 1775
ADR: "8",II
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty