Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-(Trifluoromethyl)benzhydrol 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Phenyl(4-(trifluoromethyl)phenyl)methanol

L14147-06 L14147-03 L14147-06 L14147-03
AAAL14147-06EA 152.99 USD InStock
AAAL14147-06 CAAAAL14147-03 CAAAAL14147-06 AAAL14147-03
4-(Trifluoromethyl)benzhydrol 97%
4-(Trifluoromethyl)benzhydrol


Formula: C₁₄H₁₁F₃O
MW: 252.23 g/mol
Boiling Pt: 122…124 °C/0.4 mm
Melting Pt: 59…61 °C
Storage Temperature: Ambient
MDL Number: MFCD00014399
CAS Number: 395-23-3
EINECS: 209-917-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty