Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(R)-(-)-2-Amino-3-methyl-1-butanol 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (R)-(-)-2-Amino-3-methylbutanolD(-)-2-Amino-3-methyl-1-butanolD(-)-ValinolD-Valinol

L14166-03 L14166-06 L14166-06 L14166-03
AAAL14166-06EA 190.99 USD InStock
AAAL14166-06 CAAAAL14166-03 AAAL14166-03 CAAAAL14166-06
(R)-(-)-2-Amino-3-methyl-1-butanol 98%
(R)-(-)-2-Amino-3-methyl-1-butanol


Formula: C₅H₁₃NO
MW: 103.16 g/mol
Boiling Pt: 189…190 °C
Melting Pt: 30…34 °C
Density: 0.931
Flash Pt: 78 °C(172 °F)
Storage Temperature: Ambient
MDL Number: MFCD00064297
CAS Number: 4276-09-9
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty