Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-N-Methylglucamine 99%

Supplier: Thermo Scientific Chemicals
Synonyms: N-Methyl-D-glucamine(-)-N-Methyl-D-glucamine(-)-1-Methylamino-1-deoxy-D-glucitol(-)-1-Deoxy-1-(methylamino)-D-glucitolMeglumine(2R,3R,4R,5S)-6-(Methylamino)hexane-1,2,3,4,5-pentaol

L14282-18 L14282-30 L14282-30 L14282-18
AAAL14282-30EA 62.49 USD InStock
AAAL14282-30 CAAAAL14282-30 AAAL14282-18 CAAAAL14282-18
D-(-)-N-Methylglucamine 99%
D(-)-N-Methylglucamine


Formula: H₃CNHCH₂(CH(OH))₄CH₂OH
MW: 195.22 g/mol
Melting Pt: 128…131 °C
Storage Temperature: Ambient
MDL Number: MFCD00004707
CAS Number: 6284-40-8
EINECS: 228-506-9
Merck Index: 12,06154
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty