Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

N-Acetyl-L-proline 99%

Supplier: Thermo Scientific Chemicals
Synonyms: Ac-Pro-OH

L14300-03 L14300-06 L14300-03 L14300-06
AAAL14300-03EA 38.29 USD InStock
AAAL14300-03 CAAAAL14300-03 AAAL14300-06 CAAAAL14300-06
N-Acetyl-L-proline 99%
N-Acetyl-L-proline


Formula: C₇H₁₁NO₃
Melting Pt: 117…118 °C
Storage Temperature: Ambient
MDL Number: MFCD00020837
CAS Number: 68-95-1
EINECS: 200-698-9
Order Now
Specification Test Results
Optical rotation -116° (c=1 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty