Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: (-)-Mosher's Acid Chloride(-)-MTPA-Cl(R)-(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride

L14325-03 L14325-MF L14325-03 L14325-MD L14325-MD L14325-MF
AAAL14325-03EA 488.99 USD InStock
CAAAAL14325-03 AAAL14325-03 CAAAAL14325-MD CAAAAL14325-MF AAAL14325-MD AAAL14325-MF
(-)-ɑ-Methoxy-ɑ-(trifluoromethyl)phenylacetyl chloride ≥98%
(-)-alpha-Methoxy-alpha-(trifluoromethyl)phenylacetyl chloride


Formula: C₁₀H₈ClF₃O₂
MW: 252.62 g/mol
Boiling Pt: 213…214 °C
Density: 1.353
Flash Pt: 89 °C(192 °F)
Storage Temperature: Refrigerator
MDL Number: MFCD00044400
CAS Number: 39637-99-5
UN: 3265
ADR: "8",III
Order Now
Specification Test Results
Optical rotation -137° (c=6.4 in carbon tetrachloride)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty