Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-Glucoronic acid ≥98%

Supplier: Thermo Scientific Chemicals
Synonyms: D-(+)Glucuronic acidD-(+)-glucuronic acid(2S,3S,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid

L14350-06 L14350-14 L14350-06 L14350-14
AAAL14350-14EA 211.13 USD InStock
CAAAAL14350-14 CAAAAL14350-06 AAAL14350-14 AAAL14350-06
D-Glucoronic acid ≥98%
D-Glucoronic acid


Formula: C₆H₁₀O₇
MW: 194.139 g/mol
Melting Pt: ca 158 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00166981
CAS Number: 6556-12-3
EINECS: 229-486-4
Order Now
Specification Test Results
Optical rotation +35° (c=6 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty