Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Nujol (light oil) for IR spectrophotometry

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: Light liquid paraffinParaffin, liquidVaseline oilMineral oil

L14480-AP L14480-AP L14480-AD L14480-AD
AAAL14480-APEA 50.82 USD InStock
CAAAAL14480-AP AAAL14480-AP CAAAAL14480-AD AAAL14480-AD
Nujol (light oil) for IR spectrophotometry
Nujol


Boiling Pt: >300 °C
Melting Pt: ca …20 °C
Density: 0.838
Flash Pt: 220 °C(428 °F)
Storage Temperature: Ambient
MDL Number: MFCD00131611
CAS Number: 8012-95-1
EINECS: 232-384-2
Merck Index: 13,07093
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty