Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

6-Methoxy-2-naphthoic acid 98+%

Supplier: Thermo Scientific Chemicals
Warning

L14610-03 L14610-06 L14610-06 L14610-03
AAAL14610-06EA 123.99 USD InStock
AAAL14610-06 CAAAAL14610-06 AAAL14610-03 CAAAAL14610-03
6-Methoxy-2-naphthoic acid 98+%
6-Methoxy-2-naphthoic acid


Formula: C₁₂H₁₀O₃
MW: 202.21 g/mol
Melting Pt: 192…194 °C
Storage Temperature: Ambient
EINECS: 000-000-0
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty