Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Petroleum spirit, 40…60 °C

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Petroleum etherPetroleum benzine

L14649-AU L14649-0F L14649-0F L14649-AU
AAAL14649-AUEA 37.39 USD InStock
AAAL14649-AU CAAAAL14649-0F AAAL14649-0F CAAAAL14649-AU
Petroleum spirit, 40…60 °C
Petroleum spirit (8032-32-4)


Boiling Pt: 40…60 °C
Density: 0.660
Flash Pt: -40 °C(-40 °F)
Storage Temperature: Ambient
CAS Number: 8032-32-4
EINECS: 232-453-7
UN: 1268
ADR: "3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty