Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-6-fluorobenzaldehyde oxime 97%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: 2-Chloro-6-fluorobenzaldoxime

L14719-06 L14719-06 L14719-03 L14719-03
AAAL14719-03EA 37.69 USD InStock
CAAAAL14719-03 AAAL14719-03 CAAAAL14719-06 AAAL14719-06
2-Chloro-6-fluorobenzaldehyde oxime 97%
2-Chloro-6-fluorobenzaldehyde oxime


Formula: C₇H₅ClFNO
MW: 173.57 g/mol
Melting Pt: 125…127 °C
Storage Temperature: Ambient
MDL Number: MFCD00837704
CAS Number: 443-33-4
EINECS: 207-135-6
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty