Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

2-Chloro-3-nitrobenzoic acid 98%

Supplier: Thermo Scientific Chemicals
Warning

L14954-06 L14954-03 L14954-03 L14954-06
AAAL14954-06EA 165.99 USD InStock
AAAL14954-06 CAAAAL14954-03 AAAL14954-03 CAAAAL14954-06
2-Chloro-3-nitrobenzoic acid 98%
2-Chloro-3-nitrobenzoic acid


Formula: C₇H₄ClNO₄
MW: 201.57 g/mol
Melting Pt: 182…186 °C
Storage Temperature: Ambient
MDL Number: MFCD00007069
CAS Number: 3970-35-2
EINECS: 223-590-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty