Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Polysorbate 20

Supplier: Thermo Scientific Chemicals
Synonyms: Polyoxyethylene-20-Sorbitan MonolaurateSorbitan monolaurate, ethoxylated

L15029-AP L15029-AE L15029-AP L15029-AE
AAAL15029-AEEA 41.76 USD InStock
AAAL15029-AE CAAAAL15029-AP CAAAAL15029-AE AAAL15029-AP
Polysorbate 20
Polysorbate 20
Used for solubilizing membrane proteins during isolation and purification


Formula: C₅₈H₁₁₄O₂₆
Boiling Pt: >100 °C
Density: 1.105
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty