Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥97% free radical

Supplier: Thermo Scientific Chemicals
Synonyms: 3-Carboxy-PROXYL3-Carboxy-2,2,5,5-tetramethyl-1-pyrrolidinyloxy

L15068-MD L15068-MD L15068-MF L15068-MF
AAAL15068-MFEA 29.51 USD InStock
AAAL15068-MF CAAAAL15068-MF AAAL15068-MD CAAAAL15068-MD
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl ≥97% free radical
3-Carboxy-2,2,5,5-tetramethylpyrrolidine-1-oxyl


Formula: C₉H₁₆NO₃
MW: 186.23 g/mol
Melting Pt: ca 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003167
CAS Number: 2154-68-9
EINECS: 218-448-2
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty