Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Bis(cyclopentadienyl)zirconium hydridochloride 95%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Bis(cyclopentadienyl)zirconium chloride hydrideSchwartz's ReagentBis(cyclopentadienyl)zirconium(IV) chloride hydride

L15164-03 L15164-14 L15164-03 L15164-14 L15164-06 L15164-06
AAAL15164-03EA 41.39 USD InStock
AAAL15164-03 AAAL15164-14 AAAL15164-06 CAAAAL15164-06 CAAAAL15164-03 CAAAAL15164-14
Bis(cyclopentadienyl)zirconium hydridochloride 95%
Bis(cyclopentadienyl)zirconium hydridochloride


Formula: C₁₀H₁₁ClZr
MW: 257.87 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD02089401
CAS Number: 37342-97-5
EINECS: 253-479-5
UN: 3131
ADR: "4.3",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty