Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

D-(-)-Quinic acid 98%

Supplier: Thermo Scientific Chemicals
Warning
Synonyms: (1α,3R,4α,5R)-1,3,4,5-Tetrahydroxycyclohexanecarboxylic acid

L15238-06 L15238-14 L15238-06 L15238-14
AAAL15238-06EA 35.3 USD InStock
CAAAAL15238-14 AAAL15238-06 AAAL15238-14 CAAAAL15238-06
D-(-)-Quinic acid 98%
D(-)-Quinic acid


Formula: C₇H₁₂O₆
MW: 192.17 g/mol
Melting Pt: 162…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00003864
CAS Number: 77-95-2
EINECS: 201-072-8
Order Now
Specification Test Results
Optical rotation -44° (c=20 in water)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty