Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Phosphorus pentoxide - methanesulfonic acid (∼7.5% P₂O₅) (w/w)

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Eaton's Reagent

L15607-AE L15607-AE L15607-AP L15607-AP
AAAL15607-APEA 185.99 USD InStock
AAAL15607-AP CAAAAL15607-AP AAAL15607-AE CAAAAL15607-AE
Phosphorus pentoxide - methanesulfonic acid (∼7.5% P₂O₅) (w/w)
Phosphorus pentoxide - methanesulfonic acid


MW: 141.95 g/mol
Density: 1.515
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00070545
CAS Number: 39394-84-8
UN: 2922
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty