Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dodecamolybdophosphoric acid hydrate Mo ≥ 47.5%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: 12-Molybdophosphoric acid hydrateMolybdophosphoric acid hydratePhosphomolybdic acid hydrate

L15827-22 L15827-14 L15827-36 L15827-36 L15827-14 L15827-22
AAAL15827-14EA 44.49 USD InStock
CAAAAL15827-36 CAAAAL15827-14 AAAL15827-14 AAAL15827-36 CAAAAL15827-22 AAAL15827-22
Dodecamolybdophosphoric acid hydrate Mo ≥ 47.5%
Dodecamolybdophosphoric acid hydrate


Formula: H₃[P(Mo₃O₁₀)₄]·nH₂O
MW: 1843.27 g/mol
Melting Pt: 78…90 °C
Density: 3.15
Storage Temperature: Ambient
MDL Number: MFCD00149913
CAS Number: 51429-74-4
EINECS: 234-713-5
UN: 3084
ADR: "8",II
Merck Index: 12,07498
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty