Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc trifluoromethanesulfonate 98%

Supplier: Thermo Scientific Chemicals
Danger
Synonyms: Zinc(II) trifluoromethanesulfonateZinc trifluoromethanesulphonateZinc(II) trifluoromethanesulphonateZinc trifluoromethanesulfonateZinc triflate

L15969-06 L15969-14 L15969-14 L15969-06
AAAL15969-06EA 54.87 USD InStock
AAAL15969-06 CAAAAL15969-06 AAAL15969-14 CAAAAL15969-14
Zinc trifluoromethanesulfonate 98%
Zinc trifluoromethanesulphonate


Formula: C₂F₆O₆S₂Zn
MW: 363.53 g/mol
Melting Pt: >300 °C
Storage Temperature: Ambient
MDL Number: MFCD00013229
CAS Number: 54010-75-2
EINECS: 258-922-6
UN: 3261
ADR: "8",III
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty